3-(N-Methylaminocarbonyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Methylcarbamoyl)-5-nitrobenzeneboronic acid

OR9426-1G
APOSOR9426-1GEA 176 EUR
APOSOR9426-1G
3-(N-Methylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(N-Methylaminocarbonyl)-5-nitrobenzeneboronic acid
Formel: C₈H₉BN₂O₅
Molecular Weight: 223,978 g/mol
Schmelzpunkt: 219…222 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783869
CAS-Nummer: 871332-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR