6-Methoxy-imidazo[1,2-b]pyridazine 95%

Lieferant: Apollo Scientific

OR942662-1G OR942662-250MG
APOSOR942662-1GEA 734 EUR
APOSOR942662-1G APOSOR942662-250MG
6-Methoxy-imidazo[1,2-b]pyridazine 95%
6-Methoxy-imidazo[1,2-b]pyridazine
Formel: C₇H₇N₃O
Molecular Weight: 149.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12913581
CAS-Nummer: 17240-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR