6-Chloro-2-methylbenzoxazole 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-methylbenzo[d]oxazole

OR942672-1G
APOSOR942672-1GEA 65.4 EUR
APOSOR942672-1G
6-Chloro-2-methylbenzoxazole 95%
6-Chloro-2-methylbenzoxazole
Formel: C₈H₆ClNO
Molecular Weight: 167.592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027438
CAS-Nummer: 63816-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR