3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

OR9427-1G
APOSOR9427-1GEA 190 EUR
APOSOR9427-1G
3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure 98%
3-Nitro-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BN₂O₅
Schmelzpunkt: 180…184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783871
CAS-Nummer: 871332-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR