2-Amino-7-methyl-7-azaspiro[3.5]nonane 97%

Lieferant: Apollo Scientific
Achtung

OR942733-250MG
APOSOR942733-250MGEA 473 EUR
APOSOR942733-250MG
2-Amino-7-methyl-7-azaspiro[3.5]nonane 97%
2-Amino-7-methyl-7-azaspiro[3.5]nonane
Formel: C₉H₁₈N₂
Molecular Weight: 154,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198555
CAS-Nummer: 1160247-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR