3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (3-Nitro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid

OR9428-1G
APOSOR9428-1GEA 190 EUR
APOSOR9428-1G
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure 98%
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BN₂O₅
Molecular Weight: 264,0400 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783873
CAS-Nummer: 871332-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR