Ethyl 5-(cyanomethyl)furan-2-carboxylate 95%

Lieferant: Apollo Scientific

OR942839-1G
APOSOR942839-1GEA 81.2 EUR
APOSOR942839-1G
Ethyl 5-(cyanomethyl)furan-2-carboxylate 95%
Ethyl 5-(cyanomethyl)furan-2-carboxylate
Formel: C₉H₉NO₃
Molecular Weight: 179,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12403566
CAS-Nummer: 51129-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR