3-(Butylaminocarbonyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Butylcarbamoyl)-5-nitrobenzeneboronic acid

OR9429-1G
APOSOR9429-1GEA 176 EUR
APOSOR9429-1G
3-(Butylaminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(Butylaminocarbonyl)-5-nitrobenzeneboronic acid
Formel: C₁₁H₁₅BN₂O₅
Molecular Weight: 266,058 g/mol
Schmelzpunkt: 190…192 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783881
CAS-Nummer: 871332-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR