Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%

Lieferant: Apollo Scientific

OR942966-1G
APOSOR942966-1GEA 647 EUR
APOSOR942966-1G
Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%
Methyl 2-(1-methylpiperidin-4-ylidene)acetate
Formel: C₉H₁₅NO₂
Molecular Weight: 169,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756506
CAS-Nummer: 154594-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR