4-Nitro-1-naphthol 95%

Lieferant: Apollo Scientific

OR942971-1G OR942971-5G
APOSOR942971-1GEA 0 EUR
APOSOR942971-1G APOSOR942971-5G
4-Nitro-1-naphthol 95%
4-Nitro-1-naphthol
Formel: C₁₀H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02179392
CAS-Nummer: 605-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR