cis-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol

Lieferant: Apollo Scientific

Synonyme: cis-3-((tert-Butyldimethylsilyl)oxy)cyclobutanol

OR943007-1G
APOSOR943007-1GEA 807 EUR
APOSOR943007-1G
cis-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
cis-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
Formel: C₁₀H₂₂O₂Si
Molecular Weight: 202,366 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105930
CAS-Nummer: 1408074-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR