2,2-Dimethyl-3-oxocyclobutane-1-carboxylic acid

Lieferant: Apollo Scientific

OR943021-1G
APOSOR943021-1GEA 1150 EUR
APOSOR943021-1G
2,2-Dimethyl-3-oxocyclobutane-1-carboxylic acid
2,2-Dimethyl-3-oxocyclobutane-1-carboxylic acid
Formel: C₇H₁₀O₃
Molecular Weight: 142,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106160
CAS-Nummer: 3183-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR