6-Bromo-N-methylpyridin-3-amine 95%
Lieferant: Apollo Scientific
OR943102-5G
APOSOR943102-5GEA
119
EUR
APOSOR943102-5G
6-Bromo-N-methylpyridin-3-amine 95%
6-Bromo-N-methylpyridin-3-amine
Formel:
C₆H₇BrN₂ Molecular Weight: 187.037 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234191 CAS-Nummer: 1022641-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...