8-Chloroquinolin-6-ol 97%

Lieferant: Apollo Scientific

OR943283-1G
APOSOR943283-1GEA 43.5 EUR
APOSOR943283-1G
8-Chloroquinolin-6-ol 97%
8-Chloroquinolin-6-ol
Formel: C₉H₆ClNO
Molecular Weight: 179,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09951943
CAS-Nummer: 18119-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR