2-Amino-4,6-dimethylpyrimidine-5-carbonitrile 95%

Lieferant: Apollo Scientific

OR943315-1G
APOSOR943315-1GEA 806 EUR
APOSOR943315-1G
2-Amino-4,6-dimethylpyrimidine-5-carbonitrile 95%
2-Amino-4,6-dimethylpyrimidine-5-carbonitrile
Formel: C₇H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18821968
CAS-Nummer: 16341-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR