Methyl (4-aminophenoxy)acetate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(4-aminophenoxy)acetate

OR943322-1G
APOSOR943322-1GEA 142 EUR
APOSOR943322-1G
Methyl (4-aminophenoxy)acetate 95%
Methyl (4-aminophenoxy)acetate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03942351
CAS-Nummer: 59954-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR