1-Ethyl-5-nitro-1H-benzo[d]imidazole 95%

Lieferant: Apollo Scientific

OR943336-1G
APOSOR943336-1GEA 0 EUR
APOSOR943336-1G
1-Ethyl-5-nitro-1H-benzo[d]imidazole 95%
1-Ethyl-5-nitro-1H-benzo[d]imidazole
Formel: C₉H₉N₃O₂
Molecular Weight: 191,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00179555
CAS-Nummer: 90349-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR