3-(Diethylcarbamoyl)-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-(Diethylcarbamoyl)-5-nitrophenyl)boronic acid

OR9434-1G
APOSOR9434-1GEA 176 EUR
APOSOR9434-1G
3-(Diethylcarbamoyl)-5-nitrobenzeneboronic acid 98%
3-(Diethylcarbamoyl)-5-nitrobenzeneboronic acid
Formel: C₁₁H₁₅BN₂O₅
Molecular Weight: 266,058 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783874
CAS-Nummer: 871332-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR