Benzyl n-cyano carbamate 95%

Lieferant: Apollo Scientific

OR943408-1G
APOSOR943408-1GEA 415 EUR
APOSOR943408-1G
Benzyl n-cyano carbamate 95%
Benzyl n-cyano carbamate
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18206011
CAS-Nummer: 86554-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR