3-Oxocyclobutyl pivalate 95%

Lieferant: Apollo Scientific

OR943437-1G
APOSOR943437-1GEA 647 EUR
APOSOR943437-1G
3-Oxocyclobutyl pivalate 95%
3-Oxocyclobutyl pivalate
Formel: C₉H₁₄O₃
Molecular Weight: 170.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24690600
CAS-Nummer: 1071194-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR