N-Methyl-1,3-benzothiazol-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 2-methylaminobenzethiazole

OR943526-1G
APOSOR943526-1GEA 574 EUR
APOSOR943526-1G
N-Methyl-1,3-benzothiazol-2-amine 95%
N-Methyl-1,3-benzothiazol-2-amine
Formel: C₈H₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00035877
CAS-Nummer: 16954-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR