(4-Chloropyrazol-1-yl)acetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Chloro-1H-pyrazol-1-yl)acetic acid

OR943527-5G
APOSOR943527-5GEA 217 EUR
APOSOR943527-5G
(4-Chloropyrazol-1-yl)acetic acid 95%
(4-Chloropyrazol-1-yl)acetic acid
Formel: C₅H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00036053
CAS-Nummer: 32089-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR