(3R)-(+)-3-(Methylamino)pyrrolidine 95%

Lieferant: Apollo Scientific

Synonyme: (3R)-(+)-3-(Methylamino)pyrrolidine dihydrochloride

OR943558-1G
APOSOR943558-1GEA 464 EUR
APOSOR943558-1G
(3R)-(+)-3-(Methylamino)pyrrolidine 95%
(3R)-(+)-3-(Methylamino)pyrrolidine
Formel: C₅H₁₂N₂
Molecular Weight: 100,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191461
CAS-Nummer: 139015-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR