1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone 95%

Lieferant: Apollo Scientific

OR943564-1G
APOSOR943564-1GEA 311 EUR
APOSOR943564-1G
1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone 95%
1-(3,5-Dimethyl-1H-pyrazol-4-yl)ethanone
Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234353
CAS-Nummer: 1123-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR