1,3-Dihydro-2-benzofuran-5-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

OR943816-250MG
APOSOR943816-250MGEA 393 EUR
APOSOR943816-250MG
1,3-Dihydro-2-benzofuran-5-carbaldehyde 95%
1,3-Dihydro-2-benzofuran-5-carbaldehyde
Formel: C₉H₈O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09842492
CAS-Nummer: 89424-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR