C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%

Lieferant: Apollo Scientific

Synonyme: (7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)methanamine

OR943848-1G OR943848-250MG
APOSOR943848-1GEA 806 EUR
APOSOR943848-1G APOSOR943848-250MG
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine
Formel: C₈H₁₀N₄
Molecular Weight: 162.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09994896
CAS-Nummer: 1020033-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR