2,2-Bis(hydroxymethyl)propionsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

OR9441-500G
APOSOR9441-500GEA 61.1 EUR
APOSOR9441-500G
2,2-Bis(hydroxymethyl)propionsäure 98%
2,2-Bis(hydroxymethyl)propionsäure
Formel: C₅H₁₀O₄
Molecular Weight: 134.13 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004199
CAS-Nummer: 4767-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR