2,2-Bis(hydroxymethyl)butansäure 97%

Lieferant: Apollo Scientific

Synonyme: 2,2-Dimethylolbutyric Acid

OR9442-100G
APOSOR9442-100GEA 56.7 EUR
APOSOR9442-100G
2,2-Bis(hydroxymethyl)butansäure 97%
2,2-Bis(hydroxymethyl)butyric acid
Formel: C₆H₁₂O₄
Molecular Weight: 148.16 g/mol
Schmelzpunkt: 112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190088
CAS-Nummer: 10097-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR