5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 3-acetyl-1H-pyrazole-5-carboxylate
OR944243-1G
APOSOR944243-1GEA
126
EUR
APOSOR944243-1G
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester 95%
5-Acetyl-2H-pyrazole-3-carboxylic acid ethyl ester
Formel:
C₈H₁₀N₂O₃ Molecular Weight: 182.177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21362351 CAS-Nummer: 37622-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...