1-(2-Methyl-2-nitropropyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR944492-1G
APOSOR944492-1GEA 0 EUR
APOSOR944492-1G
1-(2-Methyl-2-nitropropyl)pyrrolidine 95%
1-(2-Methyl-2-nitropropyl)pyrrolidine
Formel: C₈H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00960416
CAS-Nummer: 130564-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR