2-Methoxyphenylglyoxal 95%

Lieferant: Apollo Scientific

OR944511-250MG
APOSOR944511-250MGEA 201 EUR
APOSOR944511-250MG
2-Methoxyphenylglyoxal 95%
2-Methoxyphenylglyoxal
Formel: C₉H₈O₃
Molecular Weight: 164,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06800952
CAS-Nummer: 27993-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR