Methyl 2-(4-hydroxy-2-methylphenoxy)acetate 95%

Lieferant: Apollo Scientific

OR944537-1G
APOSOR944537-1GEA 647 EUR
APOSOR944537-1G
Methyl 2-(4-hydroxy-2-methylphenoxy)acetate 95%
Methyl 2-(4-hydroxy-2-methylphenoxy)acetate
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778439
CAS-Nummer: 317319-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR