Monobenzyl malonate 95%
Lieferant: Apollo Scientific
Synonyme:
3-(benzyloxy)-3-oxopropanoic acid, 2-((Benzyloxy)carbonyl)acetic acid, malonic acid monobenzyl ester
OR944575-10G
OR944575-1G
OR944575-5G
APOSOR944575-10GEA
88.6
EUR
APOSOR944575-10G
APOSOR944575-1G
APOSOR944575-5G
Monobenzyl malonate 95%
Monobenzyl malonate
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194.186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138132 CAS-Nummer: 40204-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...