Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 95%

Lieferant: Apollo Scientific

OR944579-250MG
APOSOR944579-250MGEA 249 EUR
APOSOR944579-250MG
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 95%
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Formel: C₇H₅N₃O₂
Molecular Weight: 163.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09414725
CAS-Nummer: 1086375-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR