2-Phenylcyclopropylamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Phenylcyclopropan-1-amine
OR944624-1G
OR944624-5G
APOSOR944624-1GEA
190
EUR
APOSOR944624-1G
APOSOR944624-5G
2-Phenylcyclopropylamine 97%
2-Phenylcyclopropylamine
Formel:
C₉H₁₁N Molecular Weight: 133.19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04114536 CAS-Nummer: 54-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...