Methyl-3,3-dimethoxy-2-methylpropanoate 90%

Lieferant: Apollo Scientific

OR944664-1G
APOSOR944664-1GEA 647 EUR
APOSOR944664-1G
Methyl-3,3-dimethoxy-2-methylpropanoate 90%
Methyl-3,3-dimethoxy-2-methylpropanoate
Formel: C₇H₁₄O₄
Molecular Weight: 162,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19103373
CAS-Nummer: 76526-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR