4-(Dimethoxymethyl)benzaldehyde 90%

Lieferant: Apollo Scientific

OR944922-1G
APOSOR944922-1GEA 560 EUR
APOSOR944922-1G
4-(Dimethoxymethyl)benzaldehyde 90%
4-(Dimethoxymethyl)benzaldehyde
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11553574
CAS-Nummer: 103781-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR