4'-Methoxy-3'-methylacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Methoxy-3-methylphenyl)ethanone, 3'-Methyl-4'-methoxyacetophenone

OR944970-5G
APOSOR944970-5GEA 132 EUR
APOSOR944970-5G
4'-Methoxy-3'-methylacetophenon 95%
4'-Methoxy-3'-methylacetophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01673307
CAS-Nummer: 10024-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR