2,5-Dimethyl-4-methoxybenzoic acid 95%

Lieferant: Apollo Scientific

OR944980-1G
APOSOR944980-1GEA 0 EUR
APOSOR944980-1G
2,5-Dimethyl-4-methoxybenzoic acid 95%
2,5-Dimethyl-4-methoxybenzoic acid
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01194285
CAS-Nummer: 58106-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR