Methyl 5-aminopentanoate hydrochloride 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-aminopentanoate, hydrochloride
OR944993-5G
APOSOR944993-5GEA
116
EUR
APOSOR944993-5G
Methyl 5-aminopentanoate hydrochloride 98%
Methyl 5-aminopentanoate hydrochloride
Formel:
C₆H₁₃NO₂·ClH Molecular Weight: 167.634 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13368250 CAS-Nummer: 29840-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...