4-Methylnicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methylpyridin-3-carbonsäure, 4-Methyl-3-pyridincarbonsäure
OR9450-25G
APOSOR9450-25GEA
88.6
EUR
APOSOR9450-25G
4-Methylnicotinsäure
4-Methylnicotinsäure
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol Schmelzpunkt: 214…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128869 CAS-Nummer: 3222-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...