7-Hydroxy-6,8-diiodo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR945056-1G
APOSOR945056-1GEA 2560 EUR
APOSOR945056-1G
7-Hydroxy-6,8-diiodo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%
7-Hydroxy-6,8-diiodo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Formel: C₁₁H₁₂INO₃
Molecular Weight: 333,122 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22493438
CAS-Nummer: 160080-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR