3-Ethoxy-2-(methylamino)pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
(3-Ethoxypyridin-2-yl)methanamine
OR945187-1G
APOSOR945187-1GEA
596
EUR
APOSOR945187-1G
3-Ethoxy-2-(methylamino)pyridine 95%
3-Ethoxy-2-(methylamino)pyridine
Formel:
C₈H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061116 CAS-Nummer: 912761-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...