3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR945262-1G OR945262-250MG
APOSOR945262-1GEA 877 EUR
APOSOR945262-1G APOSOR945262-250MG
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Formel: C₉H₇NO₄
Molecular Weight: 193.156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11868820
CAS-Nummer: 24132-22-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR