(S)-4-Boc-thiomorpholine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR945274-250MG
APOSOR945274-250MGEA 705 EUR
APOSOR945274-250MG
(S)-4-Boc-thiomorpholine-3-carboxylic acid
(S)-4-Boc-thiomorpholine-3-carboxylic acid
Formel: C₁₀H₁₇NO₄S
Molecular Weight: 247,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114908
CAS-Nummer: 1187929-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR