Exo-norbornylamine hydrochloride 95%

Lieferant: Apollo Scientific

OR945288-1G
APOSOR945288-1GEA 294 EUR
APOSOR945288-1G
Exo-norbornylamine hydrochloride 95%
Exo-norbornylamine hydrochloride
Formel: C₇H₁₃N·ClH
Molecular Weight: 147,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124938
CAS-Nummer: 14370-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR