1,2-Benzoxathiin-4(3H)-one 2,2-dioxide
Lieferant: Apollo Scientific
|
Achtung
|
OR9453-100MG
APOSOR9453-100MGEA
29.2
EUR
APOSOR9453-100MG
1,2-Benzoxathiin-4(3H)-one 2,2-dioxide
1,2-Benzoxathiin-4(3H)-one 2,2-dioxide
Formel:
C₈H₆O₄S Schmelzpunkt: 88…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798366 CAS-Nummer: 49670-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...