2-(Hydroxymethyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Hydroxymethyl)phenyl)boronic acid, 2-Hydroxymethylphenylboronic acid, 2-(Hydroxymethyl)benzeneboronic acid

OR9454-5G OR9454-1G OR9454-25G
APOSOR9454-5GEA 26.1 EUR
APOSOR9454-5G APOSOR9454-1G APOSOR9454-25G
2-(Hydroxymethyl)phenylboronsäure
2-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02258966
CAS-Nummer: 87199-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR