2-Acetamido-4-methyl-pent-4-enoic acid 95%

Lieferant: Apollo Scientific

OR945430-1G
APOSOR945430-1GEA 647 EUR
APOSOR945430-1G
2-Acetamido-4-methyl-pent-4-enoic acid 95%
2-Acetamido-4-methyl-pent-4-enoic acid
Formel: C₈H₁₃NO₃
Molecular Weight: 171.194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151789
CAS-Nummer: 87325-65-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR