4-(Propionylamino)benzoic acid 95%

Lieferant: Apollo Scientific

OR945554-1G
APOSOR945554-1GEA 647 EUR
APOSOR945554-1G
4-(Propionylamino)benzoic acid 95%
4-(Propionylamino)benzoic acid
Formel: C₁₀H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01365922
CAS-Nummer: 19313-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR